Assay Details
| Gene Symbol: Trmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164559.1 | NP_001158031.1 | 7 - 8 | 993 | 98 | |
| NM_001164560.1 | NP_001158032.1 | 7 - 8 | 917 | 98 | ||
| NM_198020.2 | NP_932137.2 | 7 - 8 | 1020 | 98 | ||
| GenBank mRNA | AK084775.1 | - | 7 - 8 | 959 | 98 | |
| AK155251.1 | - | 7 - 8 | 986 | 98 | ||
| AK159467.1 | - | 7 - 8 | 958 | 98 | ||
| AK170412.1 | - | 7 - 8 | 882 | 98 | ||
| AK171002.1 | - | 7 - 8 | 985 | 98 | ||
| BC027404.1 | - | 7 - 8 | 989 | 4950980 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt1
Genomic Map