Assay Details
| Gene Symbol: Inpp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024617.3 | NP_001019788.1 | 12 - 13 | 1392 | 99 | |
| NM_001297591.1 | NP_001284520.1 | 12 - 13 | 1369 | 99 | ||
| NM_001297593.1 | NP_001284522.1 | 11 - 12 | 1250 | 99 | ||
| NM_001297596.1 | NP_001284525.1 | 12 - 13 | 1369 | 99 | ||
| GenBank mRNA | AK053554.1 | - | 5 - 6 | 521 | 99 | |
| AK089068.1 | - | 7 - 8 | 623 | 99 | ||
| AK141502.1 | - | 14 - 15 | 1366 | 99 | ||
| BC064813.1 | - | 10 - 11 | 1094 | 6826283 | 99 | |
| DQ296611.1 | - | 12 - 13 | 1369 | 99 | ||
| DQ296612.1 | - | 11 - 12 | 1250 | 99 | ||
| DQ296613.1 | - | 12 - 13 | 1369 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Inpp4b
Genomic Map