Assay Details
| Gene Symbol: Inpp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024617.3 | NP_001019788.1 | 21 - 22 | 2704 | 139 | |
| NM_001297591.1 | NP_001284520.1 | 21 - 22 | 2681 | 139 | ||
| NM_001297593.1 | NP_001284522.1 | 20 - 21 | 2562 | 139 | ||
| NM_001297596.1 | NP_001284525.1 | 21 - 22 | 2681 | 139 | ||
| GenBank mRNA | AK053554.1 | - | 14 - 15 | 1833 | 139 | |
| AK089068.1 | - | 16 - 17 | 1935 | 139 | ||
| AK139397.1 | - | 14 - 15 | 2181 | 139 | ||
| AK141502.1 | - | 23 - 24 | 2678 | 139 | ||
| BC044876.1 | - | 7 - 8 | 1068 | 139 | ||
| BC064813.1 | - | 19 - 20 | 2406 | 6826283 | 139 | |
| DQ296611.1 | - | 21 - 22 | 2681 | 139 | ||
| DQ296612.1 | - | 20 - 21 | 2562 | 139 | ||
| DQ296613.1 | - | 21 - 22 | 2681 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Inpp4b
Genomic Map