Assay Details
| Gene Symbol: Inpp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024617.3 | NP_001019788.1 | 6 - 7 | 727 | 69 | |
| NM_001297591.1 | NP_001284520.1 | 5 - 6 | 653 | 69 | ||
| NM_001297596.1 | NP_001284525.1 | 5 - 6 | 653 | 69 | ||
| GenBank mRNA | AK141502.1 | - | 7 - 8 | 650 | 69 | |
| BC064813.1 | - | 4 - 5 | 429 | 6826283 | 69 | |
| DQ296611.1 | - | 5 - 6 | 653 | 69 | ||
| DQ296613.1 | - | 5 - 6 | 653 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Inpp4b
Genomic Map