Assay Details
| Gene Symbol: Inpp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089068.1 | - | 5 - 6 | 347 | 117 | |
| DQ296611.1 | - | 10 - 11 | 1093 | 117 | ||
| DQ296612.1 | - | 9 - 10 | 974 | 117 | ||
| DQ296613.1 | - | 10 - 11 | 1093 | 117 | ||
More Information
Set Membership: |
|

Inpp4b
Genomic Map