Assay Details
| Gene Symbol: Cdh13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019707.4 | NP_062681.2 | 12 - 13 | 2021 | 95 | |
| GenBank mRNA | AB022100.1 | - | 12 - 13 | 2030 | 95 | |
| AK028002.1 | - | 4 - 5 | 833 | 95 | ||
| AK032409.1 | - | 12 - 13 | 2052 | 95 | ||
| AK039438.1 | - | 12 - 13 | 2043 | 95 | ||
| AK048724.1 | - | 12 - 13 | 2021 | 95 | ||
| AK134649.1 | - | 12 - 13 | 1980 | 95 | ||
More Information
Set Membership: |
|

Cdh13
Genomic Map