Assay Details
| Gene Symbol: Cdh13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019707.4 | NP_062681.2 | 3 - 4 | 469 | 114 | |
| GenBank mRNA | AB022100.1 | - | 3 - 4 | 478 | 114 | |
| AK016527.1 | - | 4 - 5 | 1727 | 114 | ||
| AK029195.1 | - | 3 - 4 | 428 | 114 | ||
| AK034995.1 | - | 3 - 4 | 449 | 114 | ||
| AK039438.1 | - | 3 - 4 | 491 | 114 | ||
| AK045201.1 | - | 3 - 4 | 470 | 114 | ||
| AK048724.1 | - | 3 - 4 | 469 | 114 | ||
| AK085939.1 | - | 3 - 4 | 429 | 114 | ||
| AK134649.1 | - | 3 - 4 | 428 | 114 | ||
| BC021628.1 | - | 3 - 4 | 483 | 3987627 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh13
Genomic Map