Assay Details
| Gene Symbol: Usb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133954.2 | NP_598715.2 | 5 - 6 | 100 | ||
| GenBank mRNA | AK135527.1 | - | 5 - 6 | 100 | ||
| AK160901.1 | - | 5 - 6 | 100 | |||
| AK168683.1 | - | 5 - 6 | 100 | |||
| BC016418.1 | - | 5 - 6 | 643 | 4224590 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usb1
Genomic Map