Assay Details
| Gene Symbol: Cdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK166267.1 | - | 6 - 7 | 967 | 62 | |
| BC098501.1 | - | 6 - 7 | 955 | 30023851 | 62 | |
| X06115.1 | - | 6 - 7 | 907 | 62 | ||
| X06339.1 | - | 3 - 4 | 320 | 62 | ||
More Information
Set Membership: |
|

Cdh1
Genomic Map