Assay Details
| Gene Symbol: Hsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256042.1 | NP_001242971.1 | 2 - 3 | 306 | 66 | |
| NM_011939.3 | NP_036069.1 | 2 - 3 | 306 | 66 | ||
| NR_045689.1 | 2 - 3 | 306 | 66 | |||
| GenBank mRNA | AB029349.1 | - | 2 - 3 | 238 | 66 | |
| AB029350.1 | - | 2 - 3 | 233 | 66 | ||
| AF160966.1 | - | 2 - 3 | 233 | 66 | ||
| BC138130.1 | - | 2 - 3 | 306 | 8861149 | 66 | |
| BC145153.1 | - | 2 - 3 | 306 | 66 | ||
| BC145154.1 | - | 2 - 3 | 306 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsf4
Genomic Map