Assay Details
| Gene Symbol: Hsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256042.1 | NP_001242971.1 | 3 - 4 | 432 | 73 | |
| NM_001256044.1 | NP_001242973.1 | 2 - 3 | 323 | 73 | ||
| NM_011939.3 | NP_036069.1 | 3 - 4 | 432 | 73 | ||
| NR_045689.1 | 3 - 4 | 432 | 73 | |||
| GenBank mRNA | AF160966.1 | - | 3 - 4 | 359 | 73 | |
| AK148056.1 | - | 2 - 3 | 295 | 73 | ||
| BC138130.1 | - | 3 - 4 | 432 | 8861149 | 73 | |
| BC145153.1 | - | 3 - 4 | 432 | 73 | ||
| BC145154.1 | - | 3 - 4 | 432 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsf4
Genomic Map