| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800009496.mm.1 |
NM_001256042.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009496.mm.1 |
NM_001256042.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009496.mm.1 |
NM_001256044.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009496.mm.1 |
NM_001256044.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009496.mm.1 |
NM_011939.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009496.mm.1 |
NM_011939.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009496.mm.1 |
NR_045689.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009496.mm.1 |
NR_045689.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009496.mm.1 |
TC0800001209.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800009493.mm.1 |
NM_001256042.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009493.mm.1 |
NM_001256042.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009493.mm.1 |
NM_001256044.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009493.mm.1 |
NM_001256044.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009493.mm.1 |
NM_011939.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009493.mm.1 |
NM_011939.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009493.mm.1 |
NR_045689.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009493.mm.1 |
NR_045689.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009493.mm.1 |
TC0800001209.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800009494.mm.1 |
NM_001256042.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009494.mm.1 |
NM_001256042.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009494.mm.1 |
NM_001256044.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009494.mm.1 |
NM_001256044.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009494.mm.1 |
NM_011939.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009494.mm.1 |
NM_011939.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009494.mm.1 |
NR_045689.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009494.mm.1 |
NR_045689.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800009494.mm.1 |
TC0800001209.mm.1 |
MTA TC |