Assay Details
| Gene Symbol: Hsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011939.3 | NP_036069.1 | 7 - 8 | 802 | 69 | |
| GenBank mRNA | AB029350.1 | - | 7 - 8 | 729 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Hsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011939.3 | NP_036069.1 | 7 - 8 | 802 | 69 | |
| GenBank mRNA | AB029350.1 | - | 7 - 8 | 729 | 69 | |
Set Membership: |
|