Assay Details
| Gene Symbol: Fbxo31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133765.4 | NP_598526.3 | 4 - 5 | 664 | 69 | |
| GenBank mRNA | AB041586.1 | - | 4 - 5 | 513 | 69 | |
| AK009859.1 | - | 1 - 2 | 105 | 69 | ||
| AK140412.1 | - | 4 - 5 | 665 | 69 | ||
| AK163641.1 | - | 4 - 5 | 633 | 69 | ||
| AK197038.1 | - | 1 - 2 | 113 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo31
Genomic Map