Assay Details
| Gene Symbol: 4931428F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166394.1 | NP_001159866.1 | 2 - 3 | 740 | 57 | |
| NM_028888.2 | NP_083164.1 | 3 - 4 | 1137 | 57 | ||
| GenBank mRNA | AK016481.1 | - | 2 - 3 | 1075 | 57 | |
| AK136929.1 | - | 2 - 3 | 902 | 57 | ||
| BC043046.1 | - | 2 - 3 | 897 | 5703896 | 57 | |
| BC059045.1 | - | 2 - 3 | 675 | 6816353 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

4931428F04Rik
Genomic Map