Assay Details
| Gene Symbol: Nfix | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081981.2 | NP_001075450.1 | 9 - 10 | 2388 | 51 | |
| GenBank mRNA | BC090626.1 | - | 9 - 10 | 1415 | 51 | |
| S81451.1 | - | 8 - 9 | 1399 | 51 | ||
More Information
Set Membership: |
|

Nfix
Genomic Map