Assay Details
| Gene Symbol: Ist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028018.2 | NP_082294.1 | 2 - 3 | 220 | 106 | |
| GenBank mRNA | AK010262.1 | - | 2 - 3 | 190 | 106 | |
| AK050468.1 | - | 2 - 3 | 187 | 106 | ||
| AK088192.1 | - | 2 - 3 | 175 | 106 | ||
| AK152830.1 | - | 2 - 3 | 187 | 106 | ||
| AK161899.1 | - | 2 - 3 | 176 | 106 | ||
| BC017605.1 | - | 2 - 3 | 186 | 4507347 | 106 | |
| BC039052.1 | - | 2 - 3 | 163 | 2631157 | 106 | |
| BY160969.1 | - | 2 - 3 | 221 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ist1
Genomic Map