Assay Details
| Gene Symbol: Atxn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080930.1 | NP_001074399.1 | 4 | 2352 | 94 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
extracellular matrix organization
lung alveolus development
positive regulation of hematopoietic stem cell proliferation
DNA binding
RNA binding
protein binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
extracellular matrix organization
lung alveolus development
positive regulation of hematopoietic stem cell proliferation
DNA binding
RNA binding
protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158731 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158735 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158732 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158730 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158736 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158733 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158734 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P129866 | AU023044.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P129866 | AK082766.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P129866 | NM_001080930.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P129866 | AU023044.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P129866 | AK082766.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P129866 | NM_001080930.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939799 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225920 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848000 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925620 | AK042503.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925620 | AK076520.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925620 | AK085524.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925620 | NM_001033385.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843307 | AJ011413.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843307 | AJ457860.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843307 | AK010025.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843307 | AK050248.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843307 | AK050644.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843307 | AK131891.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843307 | AK160487.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843307 | BC024643.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843307 | BC049971.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843307 | M16111.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843307 | NM_009654.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918567 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124446 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P129866 | AU023044.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P129866 | AK082766.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P129866 | NM_001080930.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P129866 | AU023044.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P129866 | AK082766.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P129866 | NM_001080930.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1156832 | AK089170.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118092 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158732 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158736 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158735 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158734 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158733 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158730 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26158731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26158639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06981764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06981763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07316331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06981792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07316323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07316330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06894830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06981799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06981762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06772927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06981780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06981759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06772928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26158701 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581639 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435921_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455507_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445624_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581636 | - | - |
| Mouse Genome 430 2.0 Array | 1445624_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455507_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1435921_at | - | - |
| Mouse Genome 430 2.0 Array | 1455507_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1445624_at | - | - |
| Mouse Genome 430 2.0 Array | 1435921_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52335_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52335_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800022386.mm.1 | NM_001080930.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022386.mm.1 | NM_001080930.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022386.mm.1 | TC0800002910.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800022389.mm.1 | NM_001080930.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022389.mm.1 | NM_001080930.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022389.mm.1 | TC0800002910.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800022392.mm.1 | NM_001080930.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022392.mm.1 | NM_001080930.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022392.mm.1 | TC0800002910.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800022394.mm.1 | NM_001080930.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022394.mm.1 | NM_001080930.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022394.mm.1 | TC0800002910.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800022387.mm.1 | NM_001080930.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022387.mm.1 | NM_001080930.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022387.mm.1 | TC0800002910.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430B Array | 1445624_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435921_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455507_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445624_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455507_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435921_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 504143 | - | - |
| ABI Mouse Genome Survey Microarray | 504723 | - | - |
| ABI Mouse Genome Survey Microarray | 659754 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2533287 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259792 | - | - |

Atxn1l
Genomic Map