Assay Details
| Gene Symbol: Crispld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030209.3 | NP_084485.1 | 1 - 2 | 135 | 65 | |
| GenBank mRNA | AK033858.1 | - | 1 - 2 | 96 | 65 | |
| AK132171.1 | - | 1 - 2 | 78 | 65 | ||
| AK149909.1 | - | 1 - 2 | 91 | 65 | ||
| AK149925.1 | - | 1 - 2 | 91 | 65 | ||
| AK150180.1 | - | 1 - 2 | 91 | 65 | ||
| AK154308.1 | - | 1 - 2 | 135 | 65 | ||
| BC090642.1 | - | 1 - 2 | 129 | 30653388 | 65 | |
| CN527154.1 | - | 1 - 2 | 129 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033858.1, AK154308.1, BC090642.1, NM_030209.3 transcripts.
Set Membership: |
|

Crispld2
Genomic Map