Assay Details
| Gene Symbol: Mon1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048143.2 | NP_001041608.1 | 4 - 5 | 1447 | 85 | |
| NM_173015.3 | NP_766603.1 | 3 - 4 | 1637 | 85 | ||
| GenBank mRNA | AK030288.1 | - | 3 - 4 | 1603 | 85 | |
| AK157390.1 | - | 4 - 5 | 1450 | 85 | ||
| AK173047.1 | - | 3 - 4 | 1309 | 85 | ||
| BC062639.1 | - | 3 - 4 | 1313 | 85 | ||
| BQ831297.1 | - | 1 - 2 | 50 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mon1b
Genomic Map