Assay Details
| Gene Symbol: Lpcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173014.1 | NP_766602.1 | 12 - 13 | 1416 | 72 | |
| GenBank mRNA | AB244716.1 | - | 12 - 13 | 1311 | 72 | |
| AK039431.1 | - | 12 - 13 | 1416 | 72 | ||
| BC144805.1 | - | 11 - 12 | 1310 | 72 | ||
| BC144806.1 | - | 10 - 11 | 1251 | 9053354 | 72 | |
| BC150666.1 | - | 10 - 11 | 1251 | 9087577 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lpcat2
Genomic Map