Assay Details
| Gene Symbol: Lpcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173014.1 | NP_766602.1 | 4 - 5 | 743 | 117 | |
| GenBank mRNA | AB244716.1 | - | 4 - 5 | 638 | 117 | |
| AK039431.1 | - | 4 - 5 | 743 | 117 | ||
| AK183634.1 | - | 2 - 3 | 137 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lpcat2
Genomic Map