Assay Details
| Gene Symbol: Lpcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC144805.1 | - | 7 - 8 | 853 | 69 | |
| BC144806.1 | - | 6 - 7 | 794 | 9053354 | 69 | |
| BC150666.1 | - | 6 - 7 | 794 | 9087577 | 69 | |
| EF442783.1 | - | 3 - 4 | 214 | 69 | ||
| EF442784.1 | - | 4 - 5 | 273 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lpcat2
Genomic Map