Assay Details
| Gene Symbol: Car7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301164.1 | NP_001288093.1 | 2 - 3 | 722 | 69 | |
| NM_001301165.1 | NP_001288094.1 | 2 - 3 | 290 | 69 | ||
| NM_053070.3 | NP_444300.1 | 2 - 3 | 317 | 69 | ||
| GenBank mRNA | AK090390.1 | - | 2 - 3 | 320 | 70 | |
| BC038270.1 | - | 2 - 3 | 697 | 69 | ||
| BC094913.1 | - | 2 - 3 | 305 | 30016726 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car7
Genomic Map