Assay Details
| Gene Symbol: Cmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163262.1 | NP_001156734.1 | 2 - 3 | 506 | 79 | |
| NM_028941.1 | NP_083217.1 | 2 - 3 | 329 | 79 | ||
| GenBank mRNA | AK016712.1 | - | 2 - 3 | 79 | ||
| AK085367.1 | - | 2 - 3 | 409 | 79 | ||
| BC158118.1 | - | 2 - 3 | 383 | 9088191 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cmip
Genomic Map