Assay Details
| Gene Symbol: Hnf4aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027970.1 | 3 - 4 | 485 | 68 | ||
| GenBank mRNA | AK002328.1 | - | 2 - 3 | 280 | 68 | |
| BC025862.1 | - | 3 - 4 | 475 | 5050923 | 68 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | NM_197983.1 | - | 475 | Chr. - - [-] on Build | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327156 | BC025862.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327156 | NR_027970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327156 | AK002328.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327156 | BC025862.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327156 | NR_027970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327156 | AK002328.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314105 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187565 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670543 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366368 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337129 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244935 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163159 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 492795 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205215 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 176834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 180487 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327156 | BC025862.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327156 | NR_027970.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327156 | AK002328.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327156 | BC025862.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327156 | NR_027970.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327156 | AK002328.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327156 | BC025862.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327156 | NR_027970.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327156 | AK002328.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327156 | BC025862.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327156 | NR_027970.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327156 | AK002328.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01523680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01514908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01523676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01523670 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10489375 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489376 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489374 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425246_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10489372 | - | - |
| Mouse Genome 430 2.0 Array | 1425246_at | - | - |
| Mouse Genome 430 2.0 Array | 1425246_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200043436.mm.1 | NR_027970.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043436.mm.1 | NR_027970.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043436.mm.1 | TC0200005092.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200043437.mm.1 | NR_027970.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043437.mm.1 | NR_027970.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043437.mm.1 | TC0200005092.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200043441.mm.1 | NR_027970.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043441.mm.1 | NR_027970.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043441.mm.1 | TC0200005092.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425246_at | - | - |
| Mouse Expression Set 430 | 1425246_PM_at | - | - |
| Murine Genome U74v2 Set | 167215_PM_at | - | - |
| Murine Genome U74v2 Set | 167215_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425246_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425246_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425246_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425246_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 355142 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5390524 | - | - |

Hnf4aos
Genomic Map