Assay Details
| Gene Symbol: Pkig | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039390.2 | NP_001034479.1 | 2 - 3 | 197 | 106 | |
| GenBank mRNA | BY266193.1 | - | 2 - 3 | 293 | 106 | |
| BY283174.1 | - | 2 - 3 | 132 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001039390.2 transcripts.
Set Membership: |
|

Pkig
Genomic Map