Assay Details
| Gene Symbol: Ttpal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029512.2 | NP_083788.2 | 2 - 3 | 564 | 110 | |
| NM_181734.3 | NP_859423.2 | 2 - 3 | 561 | 110 | ||
| GenBank mRNA | AK033600.1 | - | 2 - 3 | 510 | 110 | |
| AK039450.1 | - | 2 - 3 | 562 | 110 | ||
| AK043791.1 | - | 2 - 3 | 531 | 110 | ||
| AK052794.1 | - | 2 - 3 | 527 | 110 | ||
| AK081680.1 | - | 2 - 3 | 524 | 110 | ||
| AK156641.1 | - | 2 - 3 | 527 | 110 | ||
| BC052389.1 | - | 2 - 3 | 520 | 5718874 | 110 | |
| BC060726.1 | - | 1 - 2 | 430 | 110 | ||
| CK640285.1 | - | 1 - 2 | 197 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttpal
Genomic Map