Assay Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172760.3 | NP_766348.3 | 12 - 13 | 1304 | 66 | |
| GenBank mRNA | AK037067.1 | - | 12 - 13 | 1297 | 66 | |
| AK157875.1 | - | 1242 | 66 | |||
| BC018516.1 | - | 10 - 11 | 1193 | 3155913 | 66 | |
| BC026617.1 | - | 7 - 8 | 1249 | 66 | ||
| BC058752.1 | - | 12 - 13 | 1287 | 6485393 | 66 | |
| BC138887.1 | - | 12 - 13 | 1261 | 8861909 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elmo3
Genomic Map