Assay Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172760.3 | NP_766348.3 | 14 - 15 | 1545 | 54 | |
| GenBank mRNA | AK037067.1 | - | 14 - 15 | 1538 | 54 | |
| AK157875.1 | - | 1483 | 54 | |||
| BC018516.1 | - | 12 - 13 | 1434 | 3155913 | 54 | |
| BC026617.1 | - | 9 - 10 | 1490 | 54 | ||
| BC058752.1 | - | 14 - 15 | 1528 | 6485393 | 54 | |
| BC138887.1 | - | 14 - 15 | 1502 | 8861909 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elmo3
Genomic Map