Assay Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172760.3 | NP_766348.3 | 5 - 6 | 567 | 74 | |
| GenBank mRNA | AK037067.1 | - | 5 - 6 | 560 | 74 | |
| AK157875.1 | - | 505 | 74 | |||
| BC058752.1 | - | 5 - 6 | 550 | 6485393 | 74 | |
| BC138887.1 | - | 5 - 6 | 524 | 8861909 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elmo3
Genomic Map