Assay Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172760.3 | NP_766348.3 | 6 - 7 | 663 | 71 | |
| GenBank mRNA | AK037067.1 | - | 6 - 7 | 656 | 71 | |
| AK157875.1 | - | 601 | 71 | |||
| BC018516.1 | - | 4 - 5 | 552 | 3155913 | 71 | |
| BC026617.1 | - | 1 - 2 | 608 | 71 | ||
| BC058752.1 | - | 6 - 7 | 646 | 6485393 | 71 | |
| BC138887.1 | - | 6 - 7 | 620 | 8861909 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elmo3
Genomic Map