Assay Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172760.3 | NP_766348.3 | 7 - 8 | 820 | 72 | |
| GenBank mRNA | AK037067.1 | - | 7 - 8 | 813 | 72 | |
| AK157875.1 | - | 758 | 72 | |||
| BC018516.1 | - | 5 - 6 | 709 | 3155913 | 72 | |
| BC026617.1 | - | 2 - 3 | 765 | 72 | ||
| BC058752.1 | - | 7 - 8 | 803 | 6485393 | 72 | |
| BC138887.1 | - | 7 - 8 | 777 | 8861909 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elmo3
Genomic Map