Assay Details
| Gene Symbol: Nfatc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291168.1 | NP_001278097.1 | 1 - 2 | 190 | 67 | |
| NM_001291169.1 | NP_001278098.1 | 1 - 2 | 190 | 67 | ||
| NM_001291172.1 | NP_001278101.1 | 1 - 2 | 190 | 67 | ||
| GenBank mRNA | AK044489.1 | - | 1 - 2 | 191 | 67 | |
| EU887581.1 | - | 1 - 2 | 71 | 67 | ||
| EU887587.1 | - | 1 - 2 | 71 | 67 | ||
| EU887593.1 | - | 1 - 2 | 71 | 67 | ||
| EU887599.1 | - | 1 - 2 | 71 | 67 | ||
More Information
Set Membership: |
|

Nfatc2
Genomic Map