Assay Details
| Gene Symbol: 1500012F01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081005.1 | NP_001074474.1 | 4 - 5 | 559 | 68 | |
| GenBank mRNA | AI847202.1 | - | 1 - 2 | 68 | ||
| AK005231.1 | - | 4 - 5 | 272 | 68 | ||
| BF467288.1 | - | 4 - 5 | 559 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598309 | NM_001081005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598309 | AI847202.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598309 | AK005231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598309 | BC042795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598309 | BF467288.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598309 | NM_001081005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598309 | AI847202.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598309 | AK005231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598309 | BC042795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598309 | BF467288.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155795 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598309 | NM_001081005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598309 | AI847202.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598309 | AK005231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598309 | BC042795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598309 | BF467288.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598309 | NM_001081005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598309 | AI847202.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598309 | AK005231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598309 | BC042795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598309 | BF467288.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598309 | NM_001081005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598309 | AI847202.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598309 | AK005231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598309 | BC042795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598309 | BF467288.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598309 | NM_001081005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598309 | AI847202.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598309 | AK005231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598309 | BC042795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598309 | BF467288.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01873603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01508614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02235001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02235000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01508605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01662304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01508606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01873607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833377 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10478850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478852 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478853 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455010_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10478847 | - | - |
| Mouse Genome 430 2.0 Array | 1455010_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455010_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68949_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68949_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200021266.mm.1 | TC0200002580.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200021267.mm.1 | TC0200002580.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455010_at | - | - |
| Mouse Expression Set 430 | 1455010_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455010_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455010_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 381804 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104850204 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216409 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2939012 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2939012 | - | - |

1500012F01Rik
Genomic Map