Assay Details
| Gene Symbol: Slc9a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB089793.1 | - | 10 - 11 | 978 | 105 | |
| AF482993.1 | - | 9 - 10 | 942 | 105 | ||
| AK049410.1 | - | 10 - 11 | 974 | 105 | ||
| AK051791.1 | - | 9 - 10 | 881 | 105 | ||
| AK143281.1 | - | 10 - 11 | 947 | 105 | ||
| AK160826.1 | - | 8 - 9 | 816 | 105 | ||
| AK173069.1 | - | 10 - 11 | 1314 | 105 | ||
| BC030879.1 | - | 10 - 11 | 977 | 4241988 | 105 | |
| BC034508.1 | - | 3 - 4 | 378 | 105 | ||
| BC058947.1 | - | 10 - 11 | 948 | 5702781 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a8
Genomic Map