Assay Details
| Gene Symbol: B4galt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019835.2 | NP_062809.2 | 3 - 4 | 514 | 73 | |
| GenBank mRNA | AB004786.1 | - | 3 - 4 | 515 | 73 | |
| AF142673.1 | - | 3 - 4 | 368 | 73 | ||
| BE981379.1 | - | 3 - 4 | 384 | 73 | ||
| CJ134591.1 | - | 3 - 4 | 515 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

B4galt5
Genomic Map