Assay Details
| Gene Symbol: Elmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080287.2 | NP_525026.2 | 18 - 19 | 1900 | 107 | |
| NM_207706.1 | NP_997589.1 | 19 - 20 | 1936 | 107 | ||
| GenBank mRNA | AF398884.1 | - | 17 - 18 | 1799 | 107 | |
| AK032033.1 | - | 19 - 20 | 1909 | 107 | ||
| AK033918.1 | - | 18 - 19 | 2094 | 107 | ||
| AK035710.1 | - | 3 - 4 | 286 | 107 | ||
| AK038097.1 | - | 18 - 19 | 1889 | 107 | ||
| AK038455.1 | - | 18 - 19 | 2006 | 107 | ||
| AK045428.1 | - | 18 - 19 | 1901 | 107 | ||
| AK047040.1 | - | 18 - 19 | 1862 | 107 | ||
| AK050823.1 | - | 18 - 19 | 1863 | 107 | ||
| AK053574.1 | - | 19 - 20 | 1900 | 107 | ||
| AK129023.1 | - | 18 - 19 | 1900 | 107 | ||
| AK205725.1 | - | 1 - 2 | 93 | 107 | ||
| BC023954.1 | - | 19 - 20 | 1921 | 5320429 | 107 | |
| BC033418.1 | - | 17 - 18 | 1830 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elmo2
Genomic Map