Assay Details
| Gene Symbol: Elmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080287.2 | NP_525026.2 | 2 - 3 | 177 | 62 | |
| NM_207706.1 | NP_997589.1 | 2 - 3 | 177 | 62 | ||
| GenBank mRNA | AF398884.1 | - | 1 - 2 | 82 | 62 | |
| AK032033.1 | - | 2 - 3 | 150 | 62 | ||
| AK033918.1 | - | 2 - 3 | 371 | 62 | ||
| AK038097.1 | - | 2 - 3 | 168 | 62 | ||
| AK038455.1 | - | 2 - 3 | 283 | 62 | ||
| AK045428.1 | - | 2 - 3 | 178 | 62 | ||
| AK047040.1 | - | 2 - 3 | 139 | 62 | ||
| AK050823.1 | - | 2 - 3 | 140 | 62 | ||
| AK053574.1 | - | 2 - 3 | 141 | 62 | ||
| AK129023.1 | - | 2 - 3 | 177 | 62 | ||
| BC023954.1 | - | 2 - 3 | 162 | 5320429 | 62 | |
| BC033418.1 | - | 1 - 2 | 107 | 62 | ||
More Information
Set Membership: |
|

Elmo2
Genomic Map