Assay Details
Gene Symbol: Elmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080287.2 | NP_525026.2 | 1 - 2 | 66 | 50 | |
NM_207706.1 | NP_997589.1 | 1 - 2 | 66 | 50 | ||
GenBank mRNA | AK032033.1 | - | 1 - 2 | 39 | 50 | |
AK038097.1 | - | 1 - 2 | 57 | 50 | ||
AK045428.1 | - | 1 - 2 | 67 | 50 | ||
AK047040.1 | - | 1 - 2 | 28 | 50 | ||
AK050823.1 | - | 1 - 2 | 29 | 50 | ||
AK053574.1 | - | 1 - 2 | 30 | 50 | ||
AK129023.1 | - | 1 - 2 | 66 | 50 | ||
BC023954.1 | - | 1 - 2 | 51 | 5320429 | 50 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK032033.1, AK045428.1, AK047040.1, AK050823.1, AK053574.1, BC023954.1, NM_080287.2, NM_207706.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
