Assay Details
Gene Symbol: Dpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010072.3 | NP_034202.1 | 7 - 8 | 583 | 123 | |
GenBank mRNA | AB004789.1 | - | 7 - 8 | 557 | 123 | |
AK004834.1 | - | 7 - 8 | 576 | 123 | ||
AK005268.1 | - | 7 - 8 | 569 | 123 | ||
AK008548.1 | - | 7 - 8 | 571 | 123 | ||
BC061151.1 | - | 7 - 8 | 563 | 30307514 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
