Assay Details
Gene Symbol: Dpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010072.3 | NP_034202.1 | 4 - 5 | 397 | 156 | |
GenBank mRNA | AB004789.1 | - | 4 - 5 | 371 | 156 | |
AI099209.1 | - | 4 - 5 | 397 | 156 | ||
AK004834.1 | - | 4 - 5 | 390 | 156 | ||
AK005268.1 | - | 4 - 5 | 383 | 156 | ||
AK008548.1 | - | 4 - 5 | 156 | |||
BC061151.1 | - | 4 - 5 | 377 | 30307514 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
