Assay Details
| Gene Symbol: Adnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009628.2 | NP_033758.2 | 1 - 2 | 488 | 89 | |
| GenBank mRNA | AK129214.1 | - | 1 - 2 | 538 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_009628.2 transcripts.
Set Membership: |
|

Adnp
Genomic Map