Assay Details
| Gene Symbol: Eya2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271963.1 | NP_001258892.1 | 1 - 2 | 237 | 86 | |
| GenBank mRNA | AK137718.1 | - | 1 - 2 | 233 | 86 | |
| AK141765.1 | - | 1 - 2 | 237 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK137718.1, NM_001271963.1 transcripts.
Set Membership: |
|

Eya2
Genomic Map