Assay Details
| Gene Symbol: Nespas | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002846.2 | 1 - 2 | 370 | 72 | |||
| GenBank mRNA | AF319945.1 | - | LIT1669 Nespas |
1 - 2 | 323 | 72 | |
| AK153813.1 | - | 1 - 2 | 371 | 72 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 298 | Mir298 |
Mirn298, mir-298, mmu-mir-298 |
-18.6kb | Sense | Chr. 2 - 174267504 - 174267585 on NCBI Build GRCm38 |
| Downstream |
negative elongation factor complex member C/D, Th1l |
Nelfcd |
2410003I03Rik, C77797, Th1, Th1l |
120.4kb | Anti-sense | Chr. 2 - 174415804 - 174427503 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Nespas
Genomic Map