Assay Details
| Gene Symbol: Aurka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291185.1 | NP_001278114.1 | 3 - 4 | 393 | 79 | |
| NM_011497.4 | NP_035627.1 | 3 - 4 | 471 | 79 | ||
| GenBank mRNA | AF007817.1 | - | 3 - 4 | 418 | 79 | |
| AK077513.1 | - | 3 - 4 | 349 | 79 | ||
| AK085861.1 | - | 3 - 4 | 345 | 79 | ||
| AK145968.1 | - | 3 - 4 | 359 | 79 | ||
| AK169363.1 | - | 3 - 4 | 348 | 79 | ||
| AY336976.1 | - | 2 - 3 | 295 | 79 | ||
| BC005425.1 | - | 3 - 4 | 338 | 3590706 | 79 | |
| BC014711.1 | - | 3 - 4 | 407 | 3968835 | 79 | |
| CD560646.1 | - | 3 - 4 | 394 | 79 | ||
| U69106.1 | - | 3 - 4 | 329 | 79 | ||
| U80932.1 | - | 3 - 4 | 338 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Aurka
Genomic Map