Assay Details
| Gene Symbol: Aurka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291185.1 | NP_001278114.1 | 6 - 7 | 782 | 69 | |
| NM_011497.4 | NP_035627.1 | 6 - 7 | 860 | 69 | ||
| GenBank mRNA | AK085861.1 | - | 6 - 7 | 734 | 69 | |
| AK145968.1 | - | 6 - 7 | 748 | 69 | ||
| AK169363.1 | - | 6 - 7 | 737 | 69 | ||
| AY336976.1 | - | 5 - 6 | 684 | 69 | ||
| BC005425.1 | - | 6 - 7 | 727 | 3590706 | 69 | |
| BC014711.1 | - | 6 - 7 | 796 | 3968835 | 69 | |
| U69106.1 | - | 6 - 7 | 718 | 69 | ||
| U80932.1 | - | 6 - 7 | 727 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aurka
Genomic Map