Assay Details
| Gene Symbol: Nmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008707.3 | NP_032733.1 | 5 - 6 | 604 | 65 | |
| GenBank mRNA | AF043326.1 | - | 5 - 6 | 616 | 65 | |
| AK146523.1 | - | 5 - 6 | 619 | 65 | ||
| BC016526.1 | - | 5 - 6 | 604 | 3589678 | 65 | |
| BC021635.1 | - | 5 - 6 | 618 | 3989540 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmt1
Genomic Map