Assay Details
| Gene Symbol: Nmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008707.3 | NP_032733.1 | 6 - 7 | 725 | 67 | |
| GenBank mRNA | AF043326.1 | - | 6 - 7 | 737 | 67 | |
| AK146523.1 | - | 6 - 7 | 740 | 67 | ||
| BC016526.1 | - | 6 - 7 | 725 | 3589678 | 67 | |
| BC021635.1 | - | 6 - 7 | 739 | 3989540 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmt1
Genomic Map