Assay Details
| Gene Symbol: Plcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152813.3 | NP_690026.2 | 10 - 11 | 1826 | 60 | |
| GenBank mRNA | AK011892.1 | - | 6 - 7 | 757 | 60 | |
| AK046669.1 | - | 10 - 11 | 1792 | 60 | ||
| AK144950.1 | - | 2899 | 60 | |||
| AK173311.1 | - | 8 - 9 | 3784 | 60 | ||
| BC031392.1 | - | 10 - 11 | 1770 | 4952407 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plcd3
Genomic Map